siRNA Details
| Gene Symbol: Tcl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289468.1 | NP_001276397.1 | 1, 2 | 197 | ||
| NM_009337.3 | NP_033363.1 | 1, 2 | 197 | |||
| GenBank mRNA | AF031956.1 | - | 1, 2 | 173 | ||
| AK149361.1 | - | 1, 2 | 179 | |||
| AK163387.1 | - | 1, 2 | 179 | |||
| BC052336.1 | - | 1, 2 | 178 | 30053730 | ||
| CA558861.1 | - | 1, 2 | 166 | |||
| CJ087554.1 | - | 1, 2 | 180 | |||
| DV648028.1 | - | 1, 2 | 156 | |||
| DV653827.1 | - | 1, 2 | 197 | |||
| DV655843.1 | - | 1, 2 | 190 | |||
| Y15376.1 | - | 1, 2 | 173 | |||

Tcl1
Genomic Map