siRNA Details
| Gene Symbol: 9230019H11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK136660.1 | - | 3 | 410 | ||
| Gene Symbol: B020014A21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK139733.1 | - | 3 | 1794 | ||

9230019H11Rik
Genomic Map