siRNA Details
| Gene Symbol: Tcte2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022311.2 | NP_071706.2 | 2 | 129 | ||
| GenBank mRNA | AK005879.1 | - | 114 | |||
| AK008572.1 | - | 115 | ||||
| BC049646.1 | - | 2 | 121 | |||
| BC092528.1 | - | 2 | 129 | 6741782 | ||
| U46151.1 | - | 2 | 336 | |||

Tcte2
Genomic Map