siRNA Details
| Gene Symbol: Prss39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009355.2 | NP_033381.1 | 4 | 728 | ||
| GenBank mRNA | AB008910.1 | - | 4 | 728 | ||
| BC049616.1 | - | 4 | 723 | 6773970 | ||
| BC115665.2 | - | 4 | 727 | 40103370 | ||
| BC115666.2 | - | 4 | 727 | 40103373 | ||

Prss39
Genomic Map