siRNA Details
| Gene Symbol: Tex264 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081654.2 | NP_001075123.1 | 3 | 452 | ||
| NM_001286498.1 | NP_001273427.1 | 3 | 652 | |||
| NM_011573.3 | NP_035703.2 | 3 | 907 | |||
| GenBank mRNA | AK003113.1 | - | 3 | 415 | ||
| AK009326.1 | - | 3 | 758 | |||
| AK159046.1 | - | 2 | 1272 | |||
| AK219517.1 | - | 1 | 185 | |||
| BC002248.1 | - | 3 | 416 | 3493141 | ||
| BC006608.1 | - | 3 | 390 | 3157453 | ||
| CX220909.1 | - | 3 | 420 | |||
| DV661702.1 | - | 1 | 227 | |||

Tex264
Genomic Map