siRNA Details
| Gene Symbol: Coq6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172582.3 | NP_766170.2 | 9 | 1010 | ||
| GenBank mRNA | AK077863.1 | - | 9 | 934 | ||
| AK131889.1 | - | 9 | 972 | |||
| AK167380.1 | - | 9 | 938 | |||
| AK170311.1 | - | 8 | 1475 | |||
| BC024135.1 | - | 9 | 911 | |||
| BC051055.1 | - | 9 | 926 | |||
| BC057914.1 | - | 6 | 674 | |||
| BC088736.1 | - | 5 | 2259 | |||
| BC106183.1 | - | 9 | 994 | |||
| BC132168.1 | - | 9 | 1010 | 40130445 | ||
| BC132170.1 | - | 9 | 1010 | 40130447 | ||

Coq6
Genomic Map