siRNA Details
| Gene Symbol: Clptm1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146047.2 | NP_666159.2 | 12 | 1493 | ||
| GenBank mRNA | AK032295.1 | - | 12 | 1457 | ||
| AK048357.1 | - | 12 | 1390 | |||
| AK156203.1 | - | 12 | 1384 | |||
| AK165939.1 | - | 12 | 1354 | |||
| AK181076.1 | - | 4 | 215 | |||
| BC026562.1 | - | 12 | 1194 | 5028264 | ||

Clptm1l
Genomic Map