siRNA Details
| Gene Symbol: Clptm1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146047.2 | NP_666159.2 | 13, 14 | 1553 | ||
| GenBank mRNA | AK032295.1 | - | 13, 14 | 1517 | ||
| AK048357.1 | - | 13, 14 | 1450 | |||
| AK156203.1 | - | 13, 14 | 1444 | |||
| AK165939.1 | - | 13, 14 | 1414 | |||
| AK181076.1 | - | 5, 6 | 275 | |||
| AK187892.1 | - | 5, 6 | 259 | |||
| BC026562.1 | - | 13, 14 | 1254 | 5028264 | ||

Clptm1l
Genomic Map