siRNA Details
| Gene Symbol: Clptm1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146047.2 | NP_666159.2 | 17 | 1788 | ||
| GenBank mRNA | AK032295.1 | - | 17 | 1752 | ||
| AK048357.1 | - | 17 | 1685 | |||
| AK156203.1 | - | 17 | 1679 | |||
| AK165939.1 | - | 17 | 1649 | |||
| BC026562.1 | - | 17 | 1489 | 5028264 | ||

Clptm1l
Genomic Map