siRNA Details
| Gene Symbol: Pglyrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009402.2 | NP_033428.1 | 1 | 237 | ||
| GenBank mRNA | AF076482.1 | - | 1 | 236 | ||
| AF193843.1 | - | 1 | 225 | |||
| AK008335.1 | - | 1 | 238 | |||
| AK043227.1 | - | 1 | 235 | |||
| BC005582.1 | - | 1 | 247 | 3969014 | ||
| CT010165.1 | - | 1 | 201 | |||
| X86374.1 | - | 1 | 236 | |||

Pglyrp1
Genomic Map