siRNA Details
| Gene Symbol: Cd70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011617.2 | NP_035747.1 | 2 | 357 | ||
| GenBank mRNA | BC125407.1 | - | 2 | 316 | 40129922 | |
| BC125409.1 | - | 2 | 316 | 40129924 | ||
| U78091.1 | - | 2 | 239 | |||
| Y13636.1 | - | 2 | 357 | |||

Cd70
Genomic Map