siRNA Details
Gene Symbol: Tpm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164248.1 | NP_001157720.1 | 8 | 805 | ||
NM_001164249.1 | NP_001157721.1 | 8 | 805 | |||
NM_001164250.1 | NP_001157722.1 | 8 | 805 | |||
NM_001164251.1 | NP_001157723.1 | 8 | 805 | |||
NM_001164252.1 | NP_001157724.1 | 7 | 778 | |||
NM_001164253.1 | NP_001157725.1 | 7 | 778 | |||
NM_001164254.1 | NP_001157726.1 | 7 | 778 | |||
NM_001164255.1 | NP_001157727.1 | 8 | 805 | |||
NM_001164256.1 | NP_001157728.1 | 7 | 778 | |||
NM_024427.4 | NP_077745.2 | 8 | 805 | |||
GenBank mRNA | AK002271.1 | - | 8 | 806 | ||
AK003175.1 | - | 8 | 806 | |||
AK032942.1 | - | 8 | 854 | |||
AK077713.1 | - | 8 | 806 | |||
AK083087.1 | - | 3 | 118 | |||
AK168640.1 | - | 8 | 802 | |||
AK168783.1 | - | 8 | 802 | |||
AK169188.1 | - | 8 | 806 | |||
AK179259.1 | - | 3 | 105 | |||
AK190148.1 | - | 4 | 181 | |||
BC026720.1 | - | 8 | 779 | |||
BC132035.1 | - | 8 | 764 | 40130312 | ||
BC132037.1 | - | 8 | 764 | 40130314 | ||
BC145307.1 | - | 8 | 764 | 9053861 | ||
X64831.1 | - | 8 | 760 |
