siRNA Details
| Gene Symbol: Tnfrsf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011659.2 | NP_035789.1 | 2 | 385 | ||
| GenBank mRNA | BC065782.1 | - | 2 | 224 | ||
| BC139239.1 | - | 2 | 255 | 8862261 | ||
| BC139266.1 | - | 2 | 255 | 8862288 | ||
| Z21674.1 | - | 2 | 206 | |||

Tnfrsf4
Genomic Map