siRNA Details
| Gene Symbol: Unc119 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011676.2 | NP_035806.1 | 4, 5 | 666 | ||
| GenBank mRNA | AF030169.1 | - | 4, 5 | 690 | ||
| AK131955.1 | - | 4, 5 | 671 | |||
| BC001990.1 | - | 4, 5 | 666 | 3484396 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Unc119| Gene Symbol: Unc119 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011676.2 | NP_035806.1 | 4, 5 | 666 | ||
| GenBank mRNA | AF030169.1 | - | 4, 5 | 690 | ||
| AK131955.1 | - | 4, 5 | 671 | |||
| BC001990.1 | - | 4, 5 | 666 | 3484396 | ||