siRNA Details
Gene Symbol: Unc119 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011676.2 | NP_035806.1 | 2 | 303 | ||
GenBank mRNA | AF030169.1 | - | 2 | 327 | ||
AK131955.1 | - | 2 | 308 | |||
BC001990.1 | - | 2 | 303 | 3484396 |
