siRNA Details
| Gene Symbol: Vdac2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011695.2 | NP_035825.1 | 5 | 355 | ||
| GenBank mRNA | AK012359.1 | - | 5 | 353 | ||
| AK078555.1 | - | 5 | 359 | |||
| AK150504.1 | - | 5 | 381 | |||
| AK150627.1 | - | 5 | 381 | |||
| AK150940.1 | - | 5 | 373 | |||
| AK152274.1 | - | 5 | 353 | |||
| AK159561.1 | - | 5 | 590 | |||
| AK168199.1 | - | 5 | 326 | |||
| AK168328.1 | - | 5 | 352 | |||
| BC003731.1 | - | 5 | 355 | 3601668 | ||
| U30838.1 | - | 5 | 355 | |||

Vdac2
Genomic Map