siRNA Details
| Gene Symbol: Ccdc166 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163518.1 | NP_001156990.1 | 1 | 832 | ||
| NM_146059.2 | NP_666171.2 | 2 | 719 | |||
| GenBank mRNA | AK137409.1 | - | 1 | 532 | ||
| AK171003.1 | - | 1 | 503 | |||
| BC024536.1 | - | 1 | 440 | 4988004 | ||
| BC047213.1 | - | 1 | 833 | 6395374 | ||

Ccdc166
Genomic Map