siRNA Details
| Gene Symbol: Gxylt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033275.4 | NP_001028447.1 | 3, 4 | 498 | ||
| GenBank mRNA | AK147387.1 | - | 3, 4 | 499 | ||
| BC141302.1 | - | 3, 4 | 593 | 9055914 | ||
| BC171972.1 | - | 3, 4 | 593 | 9054258 | ||

Gxylt1
Genomic Map