siRNA Details
| Gene Symbol: Krt79 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146063.1 | NP_666175.1 | 7 | 1301 | ||
| GenBank mRNA | BC019155.1 | - | 7 | 1240 | 5065687 | |
| BC031593.1 | - | 7 | 1301 | 4935395 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krt79| Gene Symbol: Krt79 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146063.1 | NP_666175.1 | 7 | 1301 | ||
| GenBank mRNA | BC019155.1 | - | 7 | 1240 | 5065687 | |
| BC031593.1 | - | 7 | 1301 | 4935395 | ||