siRNA Details
| Gene Symbol: Ndufs8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271443.1 | NP_001258372.1 | 5 | 285 | ||
| NM_001271444.1 | NP_001258373.1 | 5 | 525 | |||
| NM_144870.5 | NP_659119.2 | 5 | 528 | |||
| GenBank mRNA | AK135069.1 | - | 5 | 286 | ||
| AK149674.1 | - | 5 | 486 | |||
| AK153182.1 | - | 5 | 472 | |||
| AK153900.1 | - | 5 | 262 | |||
| AK164901.1 | - | 5 | 262 | |||
| AK167907.1 | - | 5 | 263 | |||
| BC021616.1 | - | 5 | 246 | 5132866 | ||
| BC086766.1 | - | 5 | 239 | 6820786 | ||

Ndufs8
Genomic Map