siRNA Details
| Gene Symbol: Lrch4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168652.1 | NP_001162123.1 | 15 | 1693 | ||
| NM_146164.2 | NP_666276.1 | 15 | 1693 | |||
| GenBank mRNA | AK088331.1 | - | 15 | 1639 | ||
| AK131167.1 | - | 1853 | ||||
| AK214414.1 | - | 3 | 203 | |||
| AK217620.1 | - | 3 | 182 | |||
| BC012525.1 | - | 15 | 1633 | 2598844 | ||
| BQ924416.1 | - | 6 | 447 | |||
| CA323124.1 | - | 1 | 36 | |||
| DV650548.1 | - | 3 | 244 | |||

Lrch4
Genomic Map