siRNA Details
| Gene Symbol: Lrch4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168652.1 | NP_001162123.1 | 13 | 1464 | ||
| NM_146164.2 | NP_666276.1 | 13 | 1464 | |||
| GenBank mRNA | AK088331.1 | - | 13 | 1410 | ||
| AK131167.1 | - | 1624 | ||||
| AK190785.1 | - | 2 | 54 | |||
| AK214414.1 | - | 2 | 58 | |||
| AK217212.1 | - | 2 | 58 | |||
| BC012525.1 | - | 13 | 1404 | 2598844 | ||
| BQ924416.1 | - | 4 | 218 | |||
| DV650548.1 | - | 1 | 15 | |||

Lrch4
Genomic Map