siRNA Details
Gene Symbol: Lrch4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168652.1 | NP_001162123.1 | 14 | 1590 | ||
NM_146164.2 | NP_666276.1 | 14 | 1590 | |||
GenBank mRNA | AK088331.1 | - | 14 | 1536 | ||
AK131167.1 | - | 1750 | ||||
AK217212.1 | - | 3 | 184 | |||
AK217620.1 | - | 2 | 79 | |||
BC012525.1 | - | 14 | 1530 | 2598844 | ||
BQ924416.1 | - | 5 | 344 | |||
DV650548.1 | - | 2 | 141 |
