siRNA Details
| Gene Symbol: Adap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172723.4 | NP_766311.2 | 10 | 1100 | ||
| GenBank mRNA | AK076754.1 | - | 9 | 1086 | ||
| AK076768.1 | - | 10 | 857 | |||
| BC056934.1 | - | 9 | 826 | |||
| BC079634.1 | - | 9 | 894 | |||
| BC144874.1 | - | 9 | 842 | |||

Adap1
Genomic Map