siRNA Details
| Gene Symbol: Eva1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145570.2 | NP_663545.2 | 3 | 686 | ||
| GenBank mRNA | AK145140.1 | - | 3 | 692 | ||
| BC014699.1 | - | 3 | 619 | 4221746 | ||
| BC027150.1 | - | 3 | 641 | 4950341 | ||
| CK020834.1 | - | 3 | 607 | |||

Eva1a
Genomic Map