siRNA Details
| Gene Symbol: Ffar3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033316.2 | NP_001028488.1 | 2 | 552 | ||
| GenBank mRNA | AK148616.1 | - | 2 | 553 | ||
| BC109330.2 | - | 1 | 765 | 40051470 | ||
| BC125009.1 | - | 1 | 765 | 40051462 | ||

Ffar3
Genomic Map