siRNA Details
Gene Symbol: Mrgprb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_205795.1 | NP_991364.1 | 1 | 59 | ||
GenBank mRNA | BC107202.1 | - | 1 | 52 | ||
BC107203.1 | - | 1 | 52 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
