siRNA Details
| Gene Symbol: Adprhl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172750.3 | NP_766338.1 | 3, 4 | 630 | ||
| GenBank mRNA | AJ427360.1 | - | 3, 4 | 607 | ||
| AK081786.1 | - | 3, 4 | 464 | |||
| AK084501.1 | - | 3, 4 | 631 | |||
| BC063759.1 | - | 3, 4 | 620 | 30134000 | ||

Adprhl1
Genomic Map