siRNA Details
Gene Symbol: Naf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163564.1 | NP_001157036.1 | 8 | 1510 | ||
GenBank mRNA | AK144735.1 | - | 8 | 757 | ||
BC027148.1 | - | 4 | 293 | |||
BC053440.1 | - | 4 | 281 |
