siRNA Details
| Gene Symbol: Tusc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177709.3 | NP_808377.1 | 1 | 545 | ||
| GenBank mRNA | AK048522.1 | - | 1 | 545 | ||
| AK158395.1 | - | 1 | 508 | |||
| BC119801.1 | - | 1 | 341 | 40107649 | ||
| BC122875.1 | - | 1 | 341 | 40107655 | ||

Tusc5
Genomic Map