siRNA Details
| Gene Symbol: Fscn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172802.4 | NP_766390.2 | 1 | 256 | ||
| GenBank mRNA | AK044556.1 | - | 1 | 255 | ||
| AK050002.1 | - | 1 | 257 | |||
| AK078674.1 | - | 1 | 284 | |||
| BC109356.1 | - | 1 | 217 | 40057445 | ||
| BC109357.1 | - | 1 | 217 | 40057447 | ||

Fscn2
Genomic Map