siRNA Details
| Gene Symbol: Syt16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172804.2 | NP_766392.2 | 7 | 1725 | ||
| GenBank mRNA | AB102950.1 | - | 6 | 2057 | ||
| AK032606.1 | - | 8 | 2504 | |||
| AK137129.1 | - | 7 | 1733 | |||
| BC056932.1 | - | 6 | 1835 | 5708126 | ||

Syt16
Genomic Map