siRNA Details
Gene Symbol: Fiz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110328.1 | NP_001103798.1 | 3 | 1432 | ||
NM_001110329.1 | NP_001103799.1 | 3 | 1375 | |||
NM_001110330.1 | NP_001103800.1 | 3 | 1305 | |||
NM_011813.3 | NP_035943.3 | 3 | 1416 | |||
GenBank mRNA | AF126746.1 | - | 3 | 1366 | ||
AF126747.1 | - | 3 | 1374 | |||
AK003641.1 | - | 1 | 526 | |||
AK135672.1 | - | 3 | 1306 | |||
AK142205.1 | - | 3 | 1325 | |||
AK162454.1 | - | 3 | 1297 | |||
AK170633.1 | - | 3 | 1301 | |||
AK220212.1 | - | 3 | 1305 | |||
BC006633.1 | - | 3 | 1416 | 3482191 | ||
BC106113.1 | - | 3 | 1291 | 6492060 | ||
BC138231.1 | - | 3 | 1366 | 8861250 | ||
BC138232.1 | - | 3 | 1391 | 8861251 | ||
BC145207.1 | - | 3 | 1391 | 9053758 |
