siRNA Details
| Gene Symbol: Lynx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011838.4 | NP_035968.1 | 4 | 487 | ||
| GenBank mRNA | AF141377.1 | - | 4 | 461 | ||
| AK013827.1 | - | 4 | 484 | |||
| AK161907.1 | - | 4 | 517 | |||
| AK162920.1 | - | 4 | 510 | |||
| AK165295.1 | - | 4 | 491 | |||
| AK165314.1 | - | 4 | 491 | |||
| BC037541.1 | - | 4 | 534 | 5325123 | ||

Lynx1
Genomic Map