siRNA Details
| Gene Symbol: Mmp17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011846.4 | NP_035976.3 | 10 | 1570 | ||
| GenBank mRNA | AB021224.2 | - | 10 | 1576 | ||
| AJ010731.1 | - | 10 | 1574 | |||
| AK079221.1 | - | 1342 | ||||
| AK143521.1 | - | 10 | 1570 | |||
| AK179012.1 | - | 2 | 200 | |||
| BC051917.1 | - | 10 | 1570 | 5716458 | ||

Mmp17
Genomic Map