siRNA Details
| Gene Symbol: Nr2e3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013708.4 | NP_038736.1 | 4 | 720 | ||
| GenBank mRNA | AF148129.1 | - | 4 | 582 | ||
| AF204053.1 | - | 4 | 720 | |||
| AK044718.1 | - | 3 | 852 | |||
| AK045788.1 | - | 2 | 463 | |||
| AK045859.1 | - | 2 | 463 | |||
| AK053560.1 | - | 4 | 688 | |||
| BC017521.1 | - | 4 | 714 | 4512088 | ||
| BC047150.1 | - | 1 | 1407 | |||
| BC057958.1 | - | 1 | 1447 | |||

Nr2e3
Genomic Map