siRNA Details
| Gene Symbol: Nr2e3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013708.4 | NP_038736.1 | 8 | 1321 | ||
| GenBank mRNA | AF148129.1 | - | 8 | 1183 | ||
| AF204053.1 | - | 8 | 1321 | |||
| AK044718.1 | - | 7 | 1453 | |||
| AK045788.1 | - | 6 | 1064 | |||
| AK045859.1 | - | 6 | 1064 | |||
| AK053560.1 | - | 8 | 1289 | |||
| BC017521.1 | - | 8 | 1315 | 4512088 | ||
| BC047150.1 | - | 5 | 2008 | |||
| BC057958.1 | - | 4 | 2293 | |||

Nr2e3
Genomic Map