siRNA Details
| Gene Symbol: Nr2e3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013708.4 | NP_038736.1 | 3, 4 | 529 | ||
| GenBank mRNA | AF148129.1 | - | 3, 4 | 391 | ||
| AF204053.1 | - | 3, 4 | 529 | |||
| AK053560.1 | - | 3, 4 | 497 | |||
| BC017521.1 | - | 3, 4 | 523 | 4512088 | ||

Nr2e3
Genomic Map