siRNA Details
| Gene Symbol: Dmrt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177360.3 | NP_796334.2 | 2 | 1492 | ||
| GenBank mRNA | AF541936.1 | - | 2 | 1492 | ||
| BC052041.1 | - | 2 | 1486 | 6404988 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dmrt3| Gene Symbol: Dmrt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177360.3 | NP_796334.2 | 2 | 1492 | ||
| GenBank mRNA | AF541936.1 | - | 2 | 1492 | ||
| BC052041.1 | - | 2 | 1486 | 6404988 | ||