siRNA Details
| Gene Symbol: Arhgef4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC094608.1 | - | 1862 | |||
| Gene Symbol: C230030N03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK082264.1 | - | 2157 | |||

Arhgef4
Genomic Map