siRNA Details
| Gene Symbol: BC055111 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183182.3 | NP_899005.2 | 8 | 1032 | ||
| GenBank mRNA | BC055111.1 | - | 8 | 1032 | 6706604 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

BC055111| Gene Symbol: BC055111 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183182.3 | NP_899005.2 | 8 | 1032 | ||
| GenBank mRNA | BC055111.1 | - | 8 | 1032 | 6706604 | |