siRNA Details
| Gene Symbol: H1fx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198622.1 | NP_941024.1 | 1 | 465 | ||
| GenBank mRNA | BC048709.1 | - | 1 | 465 | 6513403 | |
| Gene Symbol: Gm5577 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK042699.1 | - | 133 | |||

H1fx
Genomic Map