siRNA Details
| Gene Symbol: H1fx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198622.1 | NP_941024.1 | 1 | 388 | ||
| GenBank mRNA | BC048709.1 | - | 1 | 388 | 6513403 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

H1fx| Gene Symbol: H1fx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198622.1 | NP_941024.1 | 1 | 388 | ||
| GenBank mRNA | BC048709.1 | - | 1 | 388 | 6513403 | |