siRNA Details
| Gene Symbol: Kcne2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134110.3 | NP_598871.1 | 2 | 471 | ||
| GenBank mRNA | AK008619.1 | - | 2 | 472 | ||
| BC022699.1 | - | 2 | 413 | 4481325 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcne2| Gene Symbol: Kcne2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134110.3 | NP_598871.1 | 2 | 471 | ||
| GenBank mRNA | AK008619.1 | - | 2 | 472 | ||
| BC022699.1 | - | 2 | 413 | 4481325 | ||