siRNA Details
Gene Symbol: Kcne2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134110.3 | NP_598871.1 | 2 | 281 | ||
GenBank mRNA | AK008619.1 | - | 2 | 282 | ||
BC022699.1 | - | 2 | 223 | 4481325 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Kcne2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134110.3 | NP_598871.1 | 2 | 281 | ||
GenBank mRNA | AK008619.1 | - | 2 | 282 | ||
BC022699.1 | - | 2 | 223 | 4481325 |