siRNA Details
Gene Symbol: Vmn1r192 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145845.1 | NP_665844.1 | 1 | 210 | ||
GenBank mRNA | BC138395.1 | - | 1 | 303 | 8861415 | |
BC138400.1 | - | 1 | 302 | 8861420 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Vmn1r192 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145845.1 | NP_665844.1 | 1 | 210 | ||
GenBank mRNA | BC138395.1 | - | 1 | 303 | 8861415 | |
BC138400.1 | - | 1 | 302 | 8861420 |