siRNA Details
| Gene Symbol: Olfr527 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011776.1 | NP_001011776.1 | 1 | 195 | ||
| GenBank mRNA | BC147213.1 | - | 1 | 216 | 9056261 | |
| BC147214.1 | - | 1 | 216 | 9056246 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr527| Gene Symbol: Olfr527 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011776.1 | NP_001011776.1 | 1 | 195 | ||
| GenBank mRNA | BC147213.1 | - | 1 | 216 | 9056261 | |
| BC147214.1 | - | 1 | 216 | 9056246 | ||