siRNA Details
Gene Symbol: Olfr727 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146319.2 | NP_666431.2 | 1 | 958 | ||
GenBank mRNA | BC132575.1 | - | 1 | 958 | 40130852 | |
BC145712.1 | - | 1 | 958 | 40131015 |
