siRNA Details
| Gene Symbol: Olfr934 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146442.1 | NP_666653.1 | 1 | 804 | ||
| GenBank mRNA | BC113124.1 | - | 1 | 803 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr934| Gene Symbol: Olfr934 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146442.1 | NP_666653.1 | 1 | 804 | ||
| GenBank mRNA | BC113124.1 | - | 1 | 803 | ||