siRNA Details
Gene Symbol: Olfr1411 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146490.1 | NP_666701.1 | 1 | 830 | ||
GenBank mRNA | AY053401.1 | - | 1 | 430 | ||
BC128013.1 | - | 1 | 834 | 40136041 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr1411 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146490.1 | NP_666701.1 | 1 | 830 | ||
GenBank mRNA | AY053401.1 | - | 1 | 430 | ||
BC128013.1 | - | 1 | 834 | 40136041 |